Skip to Content
Merck
All Photos(1)

Key Documents

RNI00062

Sigma-Aldrich

4-Methoxy-(2-iodoethynyl)benzene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$1,308.00

R$1,308.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
R$1,308.00

About This Item

Empirical Formula (Hill Notation):
C9H7IO
CAS Number:
Molecular Weight:
258.06
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

R$1,308.00


Check Cart for Availability

Request a Bulk Order

form

solid

SMILES string

IC#CC1=CC=C(OC)C=C1

InChI

1S/C9H7IO/c1-11-9-4-2-8(3-5-9)6-7-10/h2-5H,1H3

InChI key

NMOBFTCTMWINSP-UHFFFAOYSA-N

Other Notes

All AldrichCPR products, and other building blocks or screening compounds that are part of this collection, are also available as custom packaged libraries - for more details about this service or for further information on this product, please contact us at [email protected].
Please note that Sigma-Aldrich does not collect analytical data for AldrichCPR products and that all sales are final.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service