Skip to Content
Merck
All Photos(1)

Key Documents

QBD11171

Sigma-Aldrich

DOTA-tris(acid)-amido-dPEG ®23-Maleimide

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
R$12,977.00

R$12,977.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
R$12,977.00

About This Item

Empirical Formula (Hill Notation):
C75H133F6N7O37
Molecular Weight:
1838.87
UNSPSC Code:
12352106
NACRES:
NA.22

R$12,977.00


Check Cart for Availability

Request a Bulk Order

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: chelator
reagent type: cross-linking reagent

polymer architecture

shape: linear
functionality: monofunctional

shipped in

ambient

storage temp.

−20°C

Features and Benefits

DOTA-tris(acid)-amido-dPEG®₂₃-Maleimide contains a DOTA-tris(acid) metal chelator for radio-labelling and a thiol-reactive maleimide connected by a hydrophilic dPEG spacer.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service