Key Documents
PL800GN-8
Plas-Labs neoprene glove box gloves
for 1 piece glove set, size 8 (for 8 in. ports)
Select a Size
1 PAIR
R$4,080.00
R$4,080.00
Check Cart for Availability
Select a Size
Change View
1 PAIR
R$4,080.00
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
R$4,080.00
Check Cart for Availability
material
neoprene
size
8 (for 8 in. ports)
feature
reusable
packaging
pack of 1 pair
manufacturer/tradename
Plas-Labs 800GN-8
General description
Plas-Labs black neoprene rubber gloves for chemical resistance
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
Choose from one of the most recent versions:
Certificates of Analysis (COA)