Skip to Content
Merck
All Photos(2)

Key Documents

P58200

Sigma-Aldrich

syn-2-Pyridinealdoxime

≥99%

Synonym(s):

(E)-2-Pyridinecarboxaldehyde oxime, (E)-2-Pyridylaldoxime, [C(E)]-2-Pyridinecarboxaldehyde oxime, syn-2-Pyridine aldoxime, syn-Pyridin-2-aldoxime

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
R$1,365.00

R$1,365.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
R$1,365.00

About This Item

Empirical Formula (Hill Notation):
C6H6N2O
CAS Number:
Molecular Weight:
122.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$1,365.00


Check Cart for Availability

Request a Bulk Order

Assay

≥99%

form

crystals

mp

110-112 °C (lit.)

SMILES string

O\N=C\c1ccccn1

InChI

1S/C6H6N2O/c9-8-5-6-3-1-2-4-7-6/h1-5,9H/b8-5+

InChI key

MTFJSAGADRTKCI-VMPITWQZSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service