Skip to Content
Merck
All Photos(2)

Documents

P53373

Sigma-Aldrich

Propyl isocyanate

99%

Synonym(s):

1-Isocyanatopropane, 1-Propyl isocyanate, n-Propyl isocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2NCO
CAS Number:
Molecular Weight:
85.10
Beilstein:
1098489
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.394 (lit.)

bp

83-84 °C (lit.)

density

0.908 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCN=C=O

InChI

1S/C4H7NO/c1-2-3-5-4-6/h2-3H2,1H3

InChI key

OQURWGJAWSLGQG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

32.0 °F - closed cup

Flash Point(C)

0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gala Chapman et al.
Journal of fluorescence, 29(1), 293-305 (2019-01-08)
We have synthesized and characterized of a series of single and multidye copolymerized nanoparticles with large to very large Stokes shifts (100 to 255 nm) for versatile applications as standalone or multiplexed probes in biological matrices. Nanoparticles were prepared via the
Ilaria Meazzini et al.
Journal of materials chemistry. B, 8(22), 4908-4916 (2020-04-22)
Organic-inorganic core-shell nanocomposites have attracted increasing attention for applications in imaging, controlled release, biomedical scaffolds and self-healing materials. While tunable properties can readily be achieved through the selection of complementary building blocks, synergistic enhancement requires management of the core-shell interface.
Wen-Xian Li et al.
Luminescence : the journal of biological and chemical luminescence, 35(2), 250-259 (2019-11-22)
Four core-shell structured nanometre luminescent composites with different kernel sizes and different shell layer thicknesses (SiO2(500) @Eu (phen-Si)(50) , SiO2(500) @Eu (phen-Si)(15) , SiO2(250) @Eu (phen-Si)(5) and SiO2(250) @Eu (phen-Si)(10) ) were made by changing synthesis conditions. Here, initial subscript
Jiae Seo et al.
ACS applied materials & interfaces, 11(30), 27306-27317 (2019-06-27)
A flexible hard coating material displaying extreme scratch resistance and foldable flexibility was developed via the design of an organic-inorganic hybrid coating material employing an alkoxysilyl-functionalized polyrotaxane cross-linker (PRX_Si1). PRX_Si1 has a molecular necklace-like structure that can form organic-inorganic cross-linking
Mikolaj Jan Jankowski et al.
Environmental science. Processes & impacts, 19(8), 1075-1085 (2017-08-02)
Isocyanic acid (ICA) is one of the most abundant isocyanates formed during thermal decomposition of polyurethane (PUR), and other nitrogen containing polymers. Hot-work, such as flame cutting, forging, grinding, turning and welding may give rise to thermal decomposition of said

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service