Skip to Content
Merck
All Photos(1)

Key Documents

P225

Sigma-Aldrich

[2.2]Paracyclophane

97%

Synonym(s):

Tricyclo[8.2.2.24,7]hexadeca-4,6,10,12,13,15-hexaene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
R$295.00

R$295.00


Available to ship onApril 07, 2025Details


Request a Bulk Order

Select a Size

Change View
5 G
R$295.00

About This Item

Empirical Formula (Hill Notation):
C16H16
CAS Number:
Molecular Weight:
208.30
Beilstein:
1910888
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$295.00


Available to ship onApril 07, 2025Details


Request a Bulk Order

Assay

97%

form

crystals

mp

285-288 °C (lit.)

SMILES string

C1Cc2ccc(CCc3ccc1cc3)cc2

InChI

1S/C16H16/c1-2-14-4-3-13(1)9-10-15-5-7-16(8-6-15)12-11-14/h1-8H,9-12H2

InChI key

OOLUVSIJOMLOCB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guglielmo Monaco et al.
The journal of physical chemistry. A, 123(8), 1558-1569 (2019-02-02)
The pseudostagnation graphs of the current density induced by a magnetic field perpendicular to the plane of the carbon atoms of benzene and cyclopropane, chosen as archetypal of π- and σ-aromatic molecules, have been worked out. Saddle nodes forming the
Kevin Issler et al.
Faraday discussions, 212(0), 83-100 (2018-09-22)
We investigated the excited-state dynamics of para-xylylene using a combination of field-induced surface hopping (FISH) simulations and time-resolved ionisation experiments. Our simulations predict an ultrafast decay of the initially excited bright state (S2/S3) to the S1 state on a sub-100
Christian Knierim et al.
Macromolecular bioscience, 15(8), 1052-1059 (2015-04-08)
Herein, we report on composite materials of biologically active microorganisms placed in a synthetic polymer matrix. These so-called "living composites" were utilized for gold sequestration (Micrococcus luteus) and bioremediation of nitrite (Nitrobacter winogradskyi) to demonstrate functionality. For the preparation of
null
Clay O Lacefield et al.
Cell reports, 26(8), 2000-2008 (2019-02-21)
The mammalian brain can form associations between behaviorally relevant stimuli in an animal's environment. While such learning is thought to primarily involve high-order association cortex, even primary sensory areas receive long-range connections carrying information that could contribute to high-level representations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service