Skip to Content
Merck
All Photos(1)

Key Documents

NRS3HT7

Norell® Select Series 3 mm high-throughput NMR tube

L 7 in.

Synonym(s):

routein NMR tubes, single use NMR tubes

Sign Into View Organizational & Contract Pricing

Select a Size

25 EA
R$580.00

R$580.00


Estimated to ship onApril 03, 2025



Select a Size

Change View
25 EA
R$580.00

About This Item

UNSPSC Code:
41121705
NACRES:
SB.32

R$580.00


Estimated to ship onApril 03, 2025


material

borosilicate glass (ASTM Type 1 class A)
borosilicate glass tube

feature

disposable

packaging

pkg of 25 ea

manufacturer/tradename

Norell S-3-HT-7

L

7 in.

camber

0.040 mm

diam.

3 mm

tube diam.

3 mm

wall thickness

0.58 mm

Looking for similar products? Visit Product Comparison Guide

General description

The high-throughput NMR tubes are ideal for reducing salt effects when running buffered solutions for bio-samples. They are made from ASTM Type 1 Class A borosilicate glass for reproducibility and durability, and are specially designed for one time use for routine NMR.

Legal Information

Norell is a registered trademark of Norell, Inc.
Select Series is a trademark of Norell, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service