Skip to Content
Merck
All Photos(1)

Key Documents

N17734

Sigma-Aldrich

6-Nitroindoline

97%

Synonym(s):

6-Nitro-2,3-dihydroindole, NSC 80658

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8N2O2
CAS Number:
Molecular Weight:
164.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

67-69 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc2CCNc2c1

InChI

1S/C8H8N2O2/c11-10(12)7-2-1-6-3-4-9-8(6)5-7/h1-2,5,9H,3-4H2

InChI key

LTNYDSMDSLOMSM-UHFFFAOYSA-N

Application

  • Reactant for preparation of 2-[(2,3-Dihydro-1H-indol-1-yl)methyl]melatonin analogs as novel MT2-selective melatonin receptor antagonists
  • Reactant for synthesis of isoindigo and azaisoindigo glycosides as anticancer agents
  • Reactant for preparation of arylfurancarboxamides with Nav1.8 voltage-gated Na channel blocking/analgesic activities
  • Reactant for preparation of bicyclic benzamides as 5-HT1 receptor agonists
  • Reactant for preparation of antiarrhythmic benzopyrans
  • Reactant for preparation of 5-HT2C/2B receptor antagonists

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service