Skip to Content
Merck
All Photos(1)

Key Documents

M51601

Sigma-Aldrich

2-Methylindoline

98%

Synonym(s):

α-Methyldihydroindole, 2,3-Dihydro-2-methylindole, 2-Methyl-2,3-dihydroindole, NSC 65598

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11N
CAS Number:
Molecular Weight:
133.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.569 (lit.)

bp

228-229 °C (lit.)

density

1.023 g/mL at 25 °C (lit.)

SMILES string

CC1Cc2ccccc2N1

InChI

1S/C9H11N/c1-7-6-8-4-2-3-5-9(8)10-7/h2-5,7,10H,6H2,1H3

InChI key

QRWRJDVVXAXGBT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Inhibitors of NOD1-induced nuclear factor-κB activation
  • Red fluorescent dyes for organic light-emitting diodes
  • Norepinephrine reuptake inhibitors
  • MT2-selective melatonin receptor antagonists
  • Protease-activated receptor-1 antagonists
  • Diuretic agent indapamide
  • Dopamine D2/D4 receptor antagonists
  • Antitumor agents

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

200.1 °F - closed cup

Flash Point(C)

93.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service