M26402
Methyl acetoacetate
≥99%
Synonym(s):
3-Oxobutanoic acid methyl ester, Acetoacetic acid methyl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3COCH2COOCH3
CAS Number:
Molecular Weight:
116.12
Beilstein:
506727
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
Assay
≥99%
autoignition temp.
536 °F
refractive index
n20/D 1.419 (lit.)
bp
169-170 °C/70 mmHg (lit.)
mp
−80 °C (lit.)
density
1.076 g/mL at 25 °C (lit.)
SMILES string
COC(=O)CC(C)=O
InChI
1S/C5H8O3/c1-4(6)3-5(7)8-2/h3H2,1-2H3
InChI key
WRQNANDWMGAFTP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Xiao Hu et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(47), 16796-16801 (2014-11-05)
Much remains unknown regarding speciation. Host-pathogen interactions are a major driving force for diversification, but the genomic basis for speciation and host shifting remains unclear. The fungal genus Metarhizium contains species ranging from specialists with very narrow host ranges to
L Sweetman et al.
Biomedical mass spectrometry, 5(3), 198-207 (1978-03-01)
A number of previously unrecognized abnormal metabolites have been identified and quantitated in the urine of a patient with an inherited deficiency of propionyl-CoA carboxylase. These included the isoleucine metabolites 2-methyl-3-hydroxybutyric acid and 2-methylacetoacetic acid. These isomers 3-hydroxyvaleric acid and
Karl Sörelius et al.
Circulation, 130(24), 2136-2142 (2014-11-08)
Mycotic aortic aneurysm (MAA) is a rare and life-threatening disease. The aim of this European multicenter collaboration was to study the durability of endovascular aortic repair (EVAR) of MAA, by assessing late infection-related complications and long-term survival. All EVAR treated
S Aramaki et al.
Journal of inherited metabolic disease, 14(1), 63-74 (1991-01-01)
The concentrations of 2-methylacetoacetate, 2-methyl-3-hydroxybutyrate and tiglylglycine were determined by gas chromatography-mass spectrometry in urine collected before and for 8 h after loading with 100 mg of isoleucine per kg of body weight. The sum of 2-methylacetoacetate and 2-butanone, a
Marianne Ernst et al.
Applied microbiology and biotechnology, 66(6), 629-634 (2004-11-19)
A whole-cell biotransformation system for the reduction of prochiral carbonyl compounds, such as methyl acetoacetate, to chiral hydroxy acid derivatives [methyl (R)-3-hydroxy butanoate] was developed in Escherichia coli by construction of a recombinant oxidation/reduction cycle. Alcohol dehydrogenase from Lactobacillus brevis
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service