M13602
3-Methoxycinnamic acid
97%
Synonym(s):
trans-3-(3-Methoxyphenyl)acrylic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3OC6H4CH=CHCO2H
CAS Number:
Molecular Weight:
178.18
Beilstein:
2209733
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
97%
mp
116-119 °C (lit.)
SMILES string
COc1cccc(\C=C\C(O)=O)c1
InChI
1S/C10H10O3/c1-13-9-4-2-3-8(7-9)5-6-10(11)12/h2-7H,1H3,(H,11,12)/b6-5+
InChI key
LZPNXAULYJPXEH-AATRIKPKSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Cintia Pereira da Silva et al.
Food research international (Ottawa, Ont.), 116, 1076-1083 (2019-02-06)
This study investigated the phenolic profile of jatobá-do-cerrado (Hymenaea stignocarpa Mart.) extracts submitted to in vitro digestion, the inhibition of α-amylase and α-glucosidase activities; and the effect of jatobá-do-cerrado flour addition on the nutritional quality, glycemic index (GI) and acceptability
Mingzhe Sun et al.
Journal of separation science, 42(24), 3727-3737 (2019-10-19)
To circumvent the detrimental effects of large-volume injection with fixed-loop injector in modern supercritical fluid chromatography, the feasibility of performing multiple injection was investigated. By accumulating analytes from a certain number of continual small-volume injections, compounds can be concentrated on
Mario F Mirabelli et al.
Journal of the American Society for Mass Spectrometry, 28(8), 1676-1686 (2017-04-30)
We evaluated the contribution of gas-phase in-plume proton transfer reactions to the formation of protonated and deprotonated molecules in the MALDI process. A split sample holder was used to separately deposit two different samples, which avoids any mixing during sample
Min Kyung Song et al.
Journal of agricultural and food chemistry, 67(7), 2028-2035 (2019-01-31)
Caffeic acid phenethyl ester (CAPE) is an ester of a hydroxycinnamic acid (phenylpropanoid) and a phenylethanoid (2-phenylethanol; 2-PE), which has long been used in traditional medicine. Here, we synthesized 54 hydroxycinnamic acid-phenylethanoid esters by feeding 64 combinations of hydroxycinnamic acids
Sindhu Mathew et al.
Journal of food science and technology, 52(9), 5790-5798 (2015-09-08)
The free radical scavenging activity and reducing power of 16 phenolic compounds including four hydroxycinnamic acid derivatives namely ferulic acid, caffeic acid, sinapic acid and p-coumaric acid, benzoic acid and its derivatives namely protocatechuic acid, gallic acid and vanillic acid
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service