Skip to Content
Merck
All Photos(1)

Key Documents

M073000

Sigma-Aldrich

2-Cyanoethyl N,N,N′,N′-tetraisopropylphosphorodiamidite

Synonym(s):

Bis(diisopropylamino)(2-cyanoethoxy)phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
{[(CH3)2CH]2N}2POCH2CH2CN
CAS Number:
Molecular Weight:
301.41
Beilstein:
3590103
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
Pricing and availability is not currently available.

type

for DNA synthesis

product line

Proligo Reagents

Assay

≥99% (31P-NMR)

form

liquid

refractive index

n20/D 1.470 (lit.)

bp

100 °C/0.5 mmHg (lit.)

density

0.949 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

CC(C)N(C(C)C)P(OCCC#N)N(C(C)C)C(C)C

InChI

1S/C15H32N3OP/c1-12(2)17(13(3)4)20(19-11-9-10-16)18(14(5)6)15(7)8/h12-15H,9,11H2,1-8H3

InChI key

RKVHNYJPIXOHRW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Flam. Liq. 3 - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

119.3 °F - closed cup

Flash Point(C)

48.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service