Skip to Content
Merck
All Photos(1)

Key Documents

JRD0032

Sigma-Aldrich

1-p-Tolyl-1H-pyrazole-4-carbaldehyde

AldrichCPR

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
R$1,308.00

R$1,308.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

Select a Size

Change View
1 G
R$1,308.00

About This Item

Empirical Formula (Hill Notation):
C11H10N2O
CAS Number:
Molecular Weight:
186.21
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

R$1,308.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

form

solid

SMILES string

Cc1ccc(cc1)-n2cc(C=O)cn2

InChI

1S/C11H10N2O/c1-9-2-4-11(5-3-9)13-7-10(8-14)6-12-13/h2-8H,1H3

InChI key

LFWZUGUMUQRWIU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service