Skip to Content
Merck
All Photos(1)

Documents

JKA5158

Sigma-Aldrich

HO-PEG-NH2

average MN 20,000, HCl Salt, cross-linking reagent carboxyl reactive

Synonym(s):

HO-PEG-NH2, Polyethylene glycol, hydroxyl-PEG-Amine, HO-PEG-NH2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2CH2O)nCH2CH2NH2HCl
UNSPSC Code:
12162002
NACRES:
NA.23

product name

HO-PEG20K-NH2, HCl Salt, average Mn 20,000

form

solid

mol wt

average Mn 20,000

reaction suitability

reagent type: cross-linking reagent
reactivity: carboxyl reactive

polymer architecture

shape: linear
functionality: heterobifunctional

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Hydroxyl PEG Amine. NH2 group is stable and can be activated.

Application

Applications may include: bioconjugation, drug delivery, PEG hydrogel, crosslinker, and surface functionalization

Legal Information

Product of JenKem Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bosnjakovic, A., et al.
Analytica Chimica Acta, 720 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service