J1001
Julolidine
97%
Synonym(s):
2,3,6,7-Tetrahydro-1H,5H-benzo[ij]quinolizine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Quality Level
Assay
97%
mp
34-36 °C (lit.)
storage temp.
2-8°C
SMILES string
C1CN2CCCc3cccc(C1)c23
InChI
1S/C12H15N/c1-4-10-6-2-8-13-9-3-7-11(5-1)12(10)13/h1,4-5H,2-3,6-9H2
InChI key
DZFWNZJKBJOGFQ-UHFFFAOYSA-N
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 214, 7-13 (2019-02-12)
A new Schiff base derivative fluorescence-colorimetric chemosensor 2-hydroxy-5-[(2-hydroxy-1-naphthyl)methylideneamino]benzoic acid (H3L), has been designed and synthesized. H3L displayed high selectivity and sensitivity for detecting Cr3+, Cu2+, Fe3+ and Al3+ ions in DMF/H2O (v/v = 1/1) solution. When Cr3+, Cu2+ or Fe3+ ions were
Bioconjugate chemistry, 27(3), 604-615 (2016-02-02)
The impact of drug loading and distribution on higher order structure and physical stability of an interchain cysteine-based antibody drug conjugate (ADC) has been studied. An IgG1 mAb was conjugated with a cytotoxic auristatin payload following the reduction of interchain
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service