Skip to Content
Merck
All Photos(2)

Key Documents

H4530

Sigma-Aldrich

7-Hydroxyflavone

≥98%

Synonym(s):

7-Hydroxy-2-phenyl-4H-1-benzopyran-4-one, NSC 94258

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O3
CAS Number:
Molecular Weight:
238.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

mp

245-247 °C (lit.)

SMILES string

Oc1ccc2C(=O)C=C(Oc2c1)c3ccccc3

InChI

1S/C15H10O3/c16-11-6-7-12-13(17)9-14(18-15(12)8-11)10-4-2-1-3-5-10/h1-9,16H

InChI key

MQGPSCMMNJKMHQ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Reactant:
  • Acting as a biologically valuable acceptor in O-glycosidation reactions
  • Involved in synthesis of fully phosphorylated flavones for use as pancreatic cholesterol esterase inhibitors
  • For O-methylation with di-Me carbonate
  • Linked by a polymethylene chain for synthesis of α1-adrenoceptor antagonists
  • Involved in Baylis-Hillman reactions
  • Involved in phase-transfer catalyzed glucosylation for synthesis of glucosylated flavonoids

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Osamu Kagami et al.
Journal of bioscience and bioengineering, 106(2), 121-127 (2008-09-23)
A central part (amino-acid position 268-397 of 458 amino-acid residues) of the biphenyl dioxygenase large (alpha) subunit, BphA1, from Pseudomonas pseudoalcaligenes strain KF707 was exchanged with the corresponding part of BphA1 from another biphenyl-degrading bacterium, Pseudomonas putida strain KF715, to
Jianbo Xiao et al.
Molecular nutrition & food research, 54 Suppl 2, S253-S260 (2010-03-23)
Four flavones (flavone, 7-hydroxyflavone, chrysin, and baicalein) sharing the same B- and C-ring structure but a different numbers of hydroxyl groups on the A-ring were studied for their affinities for BSA and HSA. The hydroxylation on ring A of flavones
R Le Lain et al.
Journal of enzyme inhibition, 16(1), 35-45 (2001-08-11)
In a screening programme for inhibitors of human testis 17beta-hydroxysteroid dehydrogenase (17beta-HSD type 3), as potential agents for the treatment of hormone-dependent prostatic cancer, we have used crude human testis microsomal 17beta-hydroxysteroid dehydrogenase as a convenient source of the enzyme.
Nga Ta et al.
The Journal of steroid biochemistry and molecular biology, 107(1-2), 127-129 (2007-07-13)
Previous studies have shown chrysin, 7-hydroxyflavone and 7,4'-dihydroxyflavone to be the most potent flavonoid inhibitors of aromatase. However, very poor oral bioavailability is a major limitation for the successful use of dietary flavonoids as chemopreventive agents. We have recently shown
T K Vinh et al.
Journal of enzyme inhibition, 16(5), 417-424 (2002-03-28)
A series of 4-aryl substituted 7-hydroxy-flavones were prepared using the three-step Baker-Venkataraman synthesis in good overall yields. The flavones were all evaluated in vitro for inhibitory activity against aromatase (P450AROM, CYP19), using human placental microsomes, and for inhibitory activity against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service