GF74538041
Incoloy® alloy 800 - heat resisting alloy, Ni32.5Cr21Fe
1000mm tube, 9.5mm outside diameter, 0.7mm wall thickness, 8.1mm inside diameter, annealed
Synonym(s):
NI127400
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352300
NACRES:
NA.23
Recommended Products
form
tubes
manufacturer/tradename
Goodfellow 745-380-41
L × thickness
1000 mm × 0.7 mm
O.D. × I.D.
9.5 mm × 8.1 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Legal Information
INCONEL is a registered trademark of Special Metals Corporation
Incoloy is a registered trademark of Special Metals Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service