Skip to Content
Merck
All Photos(1)

Key Documents

GF73551023

Aluminum carbide

powder, 45 max. part. size (micron), weight 50 g, purity 98.%

Synonym(s):

AL516010

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
R$5,384.00

R$5,384.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
R$5,384.00

About This Item

UNSPSC Code:
12352311
NACRES:
NA.23

R$5,384.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

powder

manufacturer/tradename

Goodfellow 735-510-23

weight

50 g

max. part. size

45 (micron)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3 - Water-react 2

Target Organs

Respiratory system

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service