Skip to Content
Merck
All Photos(1)

Key Documents

GF68381905

Nickel - Nanocrystalline

foil, light tested, 25x25mm, thickness 0.05mm, 10-20nm nominal grain size, permanent copper support

Synonym(s):

Nickel - Nanocrystalline, NJ000300

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12141711
NACRES:
NA.23

form

foil

manufacturer/tradename

Goodfellow 683-819-05

size × thickness

25x25 mm × 0.05 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service