Skip to Content
Merck
All Photos(1)

Documents

GF24744775

Invar® - Controlled Expansion Alloy, Fe64Ni36

foil, 8mm disks, 0.025mm thickness

Synonym(s):

FE020225

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101712
NACRES:
NA.23

form

foil (disc)

manufacturer/tradename

Goodfellow 247-447-75

diam. × thickness

8 mm × 0.025 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Legal Information

Invar is a registered trademark of Aperam Alloys Imphy
Invar is a registered trademark of Aperam Alloys Imphy

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service