Skip to Content
Merck
All Photos(2)

Key Documents

GF12434150

Boron

monofilament, 0.080 mm diameter, length 100 m

Synonym(s):

B 005908

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
R$3,922.00

R$3,922.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
R$3,922.00

About This Item

Empirical Formula (Hill Notation):
B
CAS Number:
Molecular Weight:
10.81
MDL number:
UNSPSC Code:
12141907
PubChem Substance ID:
NACRES:
NA.23

R$3,922.00


Check Cart for Availability

Request a Bulk Order

form

monofilament

manufacturer/tradename

Goodfellow 124-341-50

resistivity

1.5E12 μΩ-cm, 20 °C

L

100 m

diameter

0.080 mm

density

2.34 g/mL at 25 °C (lit.)

SMILES string

[B]

InChI

1S/B

InChI key

ZOXJGFHDIHLPTG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chanita Boonkanon et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 224, 117351-117351 (2019-07-25)
A tapioca starch film doped with curcumin nanoparticles was successfully fabricated and applied as a novel green colorimetric sensor for detection of boron in wastewater. Curcumin nanoparticles (curn, 30-90 nm) extracted from turmeric powder were used as a green probe, while

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service