Skip to Content
Merck
All Photos(2)

Key Documents

GF10763456

Selenium

lump, 3 mm max. lump size, weight 20 g, high purity 99.999%

Synonym(s):

SE006110

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
R$2,913.00

R$2,913.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
R$2,913.00

About This Item

Empirical Formula (Hill Notation):
Se
CAS Number:
Molecular Weight:
78.96
MDL number:
UNSPSC Code:
12141910
PubChem Substance ID:
NACRES:
NA.23

R$2,913.00


Check Cart for Availability

Request a Bulk Order

description

gray

Assay

99.999%

form

lumps

manufacturer/tradename

Goodfellow 107-634-56

resistivity

1.2 μΩ-cm, 0°C

weight

20 g

max. lump size

3 mm

bp

684.9 °C (lit.)

mp

217 °C (lit.)

density

4.81 g/mL at 25 °C (lit.)

SMILES string

[Se]

InChI

1S/Se

InChI key

BUGBHKTXTAQXES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com

Legal Information

Product of Goodfellow

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 4 - STOT RE 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service