Skip to Content
Merck
All Photos(1)

Key Documents

DWK73805B13415

Kimble® Closed Top Linerless Polypropylene Screw Thread Cap

13-415, natural screw cap

Synonym(s):

73805B-13415

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
R$1,072.00

R$1,072.00


Check Cart for Availability


Select a Size

Change View
1 PKG
R$1,072.00

About This Item

UNSPSC Code:
41100000
NACRES:
NB.17

R$1,072.00


Check Cart for Availability

material

natural screw cap
red polypropylene screw cap (Push fit)

packaging

pkg of 1 packs

manufacturer/tradename

Kimble® 73805B-13415

fitting

thread

Looking for similar products? Visit Product Comparison Guide

Application

Closure solution for vials and bottles

Features and Benefits

  • Economical, one-piece construction
  • Unique design provides exceptional sealing properties
  • Autoclavable
  • Designed from ASTM Specification E982

Legal Information

KIMBLE is a registered trademark of DWK Life Sciences

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service