Skip to Content
Merck
All Photos(2)

Key Documents

DWK293033208

Tamper-Proof Screw Caps For Wide Neck Square Bottles

neck Joints: (GL 54 H)

Synonym(s):

293033208

Sign Into View Organizational & Contract Pricing

Select a Size

10 EACH
R$195.00

R$195.00


Estimated to ship onMay 29, 2025



Select a Size

Change View
10 EACH
R$195.00

About This Item

UNSPSC Code:
41100000
NACRES:
NB.43

R$195.00


Estimated to ship onMay 29, 2025


material

blue polypropylene screw cap

Joint

Joints: neck (GL 54 H)

packaging

pkg of 10 ea

manufacturer/tradename

293033208

Looking for similar products? Visit Product Comparison Guide

Application

Sterile liquid transfer between containers, Liquid sampling into bottles from larger containers, Port assemblies for spinner flasks, and cell culture vessels,

Features and Benefits

for lime soda bottle GL 54

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service