Skip to Content
Merck
All Photos(1)

Key Documents

DWK223742

WHEATON® Type I crimp top serum bottle

without cap, round clear borosilicate glass bottle, capacity (20 mL)

Synonym(s):

223742, Crimp Top Serum Bottle, WHEATON Crimp Top Bottle

Sign Into View Organizational & Contract Pricing

Select a Size

288 EA
R$4,165.00

R$4,165.00


Check Cart for Availability


Select a Size

Change View
288 EA
R$4,165.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

R$4,165.00


Check Cart for Availability

material

round clear borosilicate glass bottle

feature

without cap

packaging

case of 288 ea Shrink-wrapped partitioned modules

manufacturer/tradename

WHEATON® 223742

bottle diam. × H × W

(25 mm) (54 mm) (276 mm)

capacity

(20 mL)

Looking for similar products? Visit Product Comparison Guide

General description

Serum Bottles with exceptional wall strength and superior heat and chemical tolerances.

  • Ideal for long and short term sample storage, lyophilization and vaccine / injectable drug containers
  • Fits most lyophilization applications
  • Manufactured from WHEATON 400 borosilicate molded glass that conforms to USP Type I requirements
  • Shrink-wrapped partitioned modules reduce breakage

Features and Benefits

  • Ideal for long and short term sample storage
  • Fits most lyophillization applications
  • Select aluminum seal and stopper size by using the mouth OD dimension of the bottle or vial
  • Shrink-wrapped partitioned modules reduce breakage
  • Borosilicate glass conforms to USP Type I requirements
  • Clear bottles manufactured from WHEATON® 400 borosilicate molded glass that conforms to USP Type I requirements
  • Amber bottles manufactured from WHEATON® 500 borosilicate glass that conforms to USP Type I requirements

Legal Information

WHEATON is a registered trademark of Wheaton Industries, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service