Skip to Content
Merck
All Photos(1)

Documents

DWK218055159

Duran® Protect, Laboratory Bottle

Laboratory bottle, plastic-coated glass bottle, neck Joints: threaded (GL45), capacity 750 mL, with screw cap to drain contents, WITH SCREW-CAP AND POURING RING (PP)

Synonym(s):

218055159

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

material

plastic-coated glass bottle

Joint

Joints: threaded neck (GL45)

feature

with screw cap to drain contents WITH SCREW-CAP AND POURING RING (PP)

packaging

pkg of 10 ea

manufacturer/tradename

DURAN® 218055159

capacity

750 mL

Looking for similar products? Visit Product Comparison Guide

General description

DURAN® Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972. The protect safety-coated version of the bottles combining the best of both worlds: virtually inert USP/EP Type I borosilicate glass 3.3 glass on the inside and a protective safety plastic layer on the outside. The bottles are moulded from high-purity, clear, glass that is then coated with a clear PU plastic coating to reduce the hazards of broken glass and liquid splashes in the event of breakage. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. The protect safety coating meets the requirements of ASTM E1522, and is fully autoclavable at 121 or 134 °C.

Application

Autoclaving of media and reagents, Preparation of media and reagents, Sample collection, Culturing vessel, Storage of media and reagents, Liquid handling reservoir, Boro 3.3, plastic coated

Features and Benefits

Standardised GL-thread and matching cap system for practicularly tight sealing and simple, plastic coated Complete with blue screw cap (PP, integral lip seal) and pouring ring (PP) for drip-free pouring and clean, safe working; Easy-to-read scale with graduation marks to estimate the content; Fired-on, highly durable white ceramic print; Sizes 10 and 25 ml with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service