Skip to Content
Merck
All Photos(1)

Documents

D77206

Sigma-Aldrich

trans-3-Hexenedinitrile

97%

Synonym(s):

trans-1,4-Dicyano-2-butene, Dihydromucononitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCCH2CH=CHCH2CN
CAS Number:
Molecular Weight:
106.13
Beilstein:
1720228
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

flakes

mp

75-77 °C (lit.)

SMILES string

N#CC\C=C\CC#N

InChI

1S/C6H6N2/c7-5-3-1-2-4-6-8/h1-2H,3-4H2/b2-1+

InChI key

BSVZXPLUMFUWHW-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service