Skip to Content
Merck
All Photos(1)

Key Documents

D75955

Sigma-Aldrich

2,5-Dichlorotoluene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3Cl2
CAS Number:
Molecular Weight:
161.03
Beilstein:
1859112
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

liquid

refractive index

n20/D 1.547 (lit.)

bp

197-200 °C (lit.)

mp

4-5 °C (lit.)

density

1.254 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(Cl)ccc1Cl

InChI

1S/C7H6Cl2/c1-5-4-6(8)2-3-7(5)9/h2-4H,1H3

InChI key

KFAKZJUYBOYVKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Riquan Liao et al.
Environmental monitoring and assessment, 191(5), 309-309 (2019-04-28)
To enhance our understanding on environmental conditions of the Maowei Sea in Guangxi province, China, the concentration and distribution of 22 chlorobenzene compounds (CBs) in the surface sediment were determined by gas chromatography-mass spectrometry (GC-MS). The relationship of the sediment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service