Skip to Content
Merck
All Photos(1)

Key Documents

D208604

Sigma-Aldrich

4,5-Diphenylimidazole

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
R$375.00

R$375.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
R$375.00

About This Item

Empirical Formula (Hill Notation):
C15H12N2
CAS Number:
Molecular Weight:
220.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

R$375.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

mp

228-230 °C (lit.)

SMILES string

c1ccc(cc1)-c2nc[nH]c2-c3ccccc3

InChI

1S/C15H12N2/c1-3-7-12(8-4-1)14-15(17-11-16-14)13-9-5-2-6-10-13/h1-11H,(H,16,17)

InChI key

CPHGOBGXZQKCKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ira J Kallio-Kujala et al.
Veterinary anaesthesia and analgesia, 45(4), 405-413 (2018-06-13)
We determined the possible effects of a peripherally acting α2-adrenoceptor antagonist, MK-467, on the absorption of intramuscularly (IM) coadministered medetomidine, butorphanol and midazolam. Randomized, experimental, blinded crossover study. Six healthy Beagle dogs. Two IM treatments were administered: 1) medetomidine hydrochloride

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service