Skip to Content
Merck
All Photos(1)

Documents

D136204

Sigma-Aldrich

3,4-Dimethoxyphenethylamine

97%

Synonym(s):

Homoveratrylamine, 2-(3,4-Dimethoxyphenyl)ethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CH2CH2NH2
CAS Number:
Molecular Weight:
181.23
Beilstein:
474393
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

drug control

USDEA Schedule I

refractive index

n20/D 1.546 (lit.)

bp

188 °C/15 mmHg (lit.)

density

1.074 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(CCN)cc1OC

InChI

1S/C10H15NO2/c1-12-9-4-3-8(5-6-11)7-10(9)13-2/h3-4,7H,5-6,11H2,1-2H3

InChI key

ANOUKFYBOAKOIR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Precursor for the synthesis of isoquinolines.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

257.0 °F

Flash Point(C)

125 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Australian Journal of Chemistry, 47, 957-957 (1994)
Julia Fedotova et al.
American journal of translational research, 7(11), 2462-2473 (2016-01-26)
Novel anxiolytic medications are necessary to broaden treatment therapy. Thus, the aim of the present study was to compare the clinically effective anxiolytic, diazepam with the novel 3,4-dimethoxyphenylethylamine derivates. The novel 3,4-dimethoxyphenylethylamine derivates (PK, 0.1, 1.0, 10.0 mg/kg, i.p.) and
Organic preparations and procedures international, 25, 91-91 (1993)
Chem. Abstr., 119, 139040g-139040g (1993)
Siyu Wang et al.
Journal of agricultural and food chemistry, 65(2), 364-372 (2016-12-23)
Wolfberry or Goji berry, the fruit of Lycium barbarum, exhibits health-promoting properties that leads to an extensive study of their active components. We synthesized a set of hydroxycinnamic acid amide (HCCA) compounds, including trans-caffeic acid, trans-ferulic acid, and 3,4-dihydroxyhydrocinnamic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service