D109800
3,4-Dihydroxybenzoic acid
97%
Synonym(s):
Protocatechuic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(HO)2C6H3CO2H
CAS Number:
Molecular Weight:
154.12
Beilstein:
1448841
EC Number:
MDL number:
UNSPSC Code:
12352103
Assay
97%
mp
197-200 °C (dec.) (lit.)
SMILES string
OC(=O)c1ccc(O)c(O)c1
InChI
1S/C7H6O4/c8-5-2-1-4(7(10)11)3-6(5)9/h1-3,8-9H,(H,10,11)
InChI key
YQUVCSBJEUQKSH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Chemopreventive in several animal models of carcinogenesis. Blocks cell proliferation in the post-initiation phase.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Frédéric Martin et al.
Journal of natural products, 72(5), 852-856 (2009-04-14)
Extracts from several plants of the family Bignoniaceae from Panama were submitted to a rapid DPPH TLC test for the detection of radical-scavenging activity. The MeOH extract of the stems of Jacaranda caucana, a tree that grows from Costa Rica
R Masella et al.
Current medicinal chemistry, 19(18), 2901-2917 (2012-04-24)
Epidemiological evidence has shown that a high dietary intake of vegetables and fruit rich in polyphenols is associated with a reduction of cancer incidence and mortality from coronary heart disease. The healthy effects associated with polyphenol consumption have made the
Lei Wang et al.
Environment international, 59, 27-32 (2013-06-12)
Human exposure to p-hydroxybenzoic acid esters (parabens) is a concern, owing to adverse health effects of these compounds. Parabens are metabolized and eliminated from the human bodies within a few hours of exposure. In this study, for the first time
J Kristinsson et al.
Pharmacology & toxicology, 74(3), 174-180 (1994-03-01)
The urinary metabolites of orally administered mebeverine hydrochloride (270 mg) were studied in five healthy volunteers with the aid of gas chromatography/mass spectrometry. Mebeverine, which is an ester of veratric acid and 4-(ethyl-[2-(4-methoxyphenyl)-1-methylethyl]amino)butan-1-ol, was completely hydrolysed to the corresponding acid
Qing-Han Gao et al.
Journal of agricultural and food chemistry, 60(38), 9642-9648 (2012-09-11)
This study investigated the changes in sugars, organic acids, α-tocopherol, β-carotene, phenolic profiles, total phenolic content (TPC), and antioxidant capacity of jujube fruits after four drying treatments (sun-, oven-, microwave- and freeze-drying). Sugar, organic acid, phenolic compounds, α-tocopherol, and β-carotene
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service