All Photos(1)
About This Item
Linear Formula:
C18H8O8Br4
CAS Number:
Molecular Weight:
671.87
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
≥98% (TLC)
SMILES string
OC(=O)c1cc(Br)cc(Br)c1OC(=O)\C=C\C(=O)Oc2c(Br)cc(Br)cc2C(O)=O
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
M Samaja et al.
Artificial cells, blood substitutes, and immobilization biotechnology, 22(3), 517-524 (1994-01-01)
The xanthine/xanthine oxidase reaction produces reproducible amounts of oxygen-derived free radicals that oxidize human oxyhemoglobin (Hb). We monitored the kinetics of the oxidation of stripped Hb (sHb), purified HbA0 and alpha-alpha cross-linked Hb (HbXL99 alpha) at [Hb] in the 5
A I Alayash et al.
Archives of biochemistry and biophysics, 303(2), 332-338 (1993-06-01)
We have examined the interactions between nitric oxide (NO) and oxidized human hemoglobin, comparing the behavior of unmodified HbA0 with that of two chemically modified hemoglobins. The latter are promising red cell substitute candidates due to their lower oxygen affinity
M S Rogers et al.
Biochimica et biophysica acta, 1248(2), 135-142 (1995-04-27)
Human hemoglobin site specifically cross-linked with bis(3,5-dibromosalicyl)fumarate results in a low oxygen affinity hemoglobin-based red cell substitute (alpha-DBBF). Polymerization of alpha-DBBF by bis(maleoylglycylamide) polyethylene glycol (BMAA-PEG) yields poly alpha-DBBF which offers the added benefits of reduced renal clearance and increased
Lei Huang et al.
Artificial cells, blood substitutes, and immobilization biotechnology, 36(2), 156-165 (2008-04-26)
Hemoglobin-based blood substitute is widely studied. As the starting materials, hemoglobin (Hb) is mainly supplied from outdated human blood and animal sources. But there are many disadvantages. As the alternative source, human placenta hemoglobin (PHb) has its own advantages. We
R Motterlini et al.
Artificial cells, blood substitutes, and immobilization biotechnology, 23(3), 291-301 (1995-01-01)
The aim of the present study was to investigate the role of hemoglobin autoxidation in the induction of endothelial heme oxygenase (HO), an inducible "stress" protein which is responsible for heme catabolism. Porcine aortic endothelial cells were incubated for six
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service