CLS99447161
Pyrex® disposable culture tubes, screw cap style with white marking spot
capacity 11.5 mL
Synonym(s):
culture tube, glass tubes, test tubes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
round bottom tube
sterility
non-sterile
feature
cap: no
packaging
case of 1000
pack of 250
manufacturer/tradename
Corning 99447-161
O.D. × H
16 mm × 100 mm
capacity
11.5 mL
color
clear
fitting
thread for 15-415
suitability
suitable for (culture)
Looking for similar products? Visit Product Comparison Guide
General description
Round bottom. Tubes are made of borosilicate glass. Screw-cap tubes are supplied without caps. Designed for both tissue culture and bacteriological work.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service