Skip to Content
Merck
All Photos(1)

Key Documents

CLS985025

Pyrex® flat bottom culture tube, rimless

capacity 39.0 mL

Synonym(s):

culture tubes, test tubes, Corning® 9850

Sign Into View Organizational & Contract Pricing

Select a Size

72 EA
R$951.00

R$951.00


Check Cart for Availability


Select a Size

Change View
72 EA
R$951.00

About This Item

UNSPSC Code:
41121706
NACRES:
NB.43

R$951.00


Check Cart for Availability

material

borosilicate glass
flat bottom tube

sterility

non-sterile

feature

cap: no

packaging

pack of 36 ea
case of 72

manufacturer/tradename

Corning 9850-25

capacity

39.0 mL

diam. × L

25 mm × 100 mm

color

clear

suitability

suitable for (plant culture)

Looking for similar products? Visit Product Comparison Guide

General description

Ideal for plant tissue culture and general culture work. The flat bottom facilitates use with agar slants.

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service