Skip to Content
Merck
All Photos(1)

Key Documents

CLS77227

Pyrex® tissue grinder

capacity 7 mL, body O.D. × H 15 mm × 117 mm

Synonym(s):

Corning® 7722

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
R$1,024.00

R$1,024.00


Check Cart for Availability


Select a Size

Change View
1 EA
R$1,024.00

About This Item

UNSPSC Code:
41121500
NACRES:
NB.43

R$1,024.00


Check Cart for Availability

manufacturer/tradename

Corning 7722-7

body O.D. × H

15 mm × 117 mm

capacity

7 mL

Related Categories

General description

Supplied with two pestles to ensure dissociation of cells into fine particles with minimal damage to cell nuclei. Use the large clearance pestle for initial reduction of soft tissue. Complete the homogenization with the small clearance pestle. Particularly useful in enzyme studies where heat build-up must be avoided.

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service