Key Documents
CLS70582X
Pyrex® disposable graduated bacteriological pipette, sterile
volume 2.2 mL
Synonym(s):
pipet
Select a Size
500 EA
R$2,683.00
R$2,683.00
Check Cart for Availability
Select a Size
Change View
500 EA
R$2,683.00
About This Item
UNSPSC Code:
41121500
NACRES:
NB.43
R$2,683.00
Check Cart for Availability
material
Pyrex
description
plugged
manufacturer/tradename
Corning 7058-2X
volume measuring range
2.2 mL
capacity
2.2 mL
General description
This pipet is very convenient when duplicate platings are to be made on the same medium or for plating differential medium for the determination of gram-negative bacteria, mold and yeast, or total plate count. Both sizes are designed for gravity feed (blow-out is not required).
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)