Key Documents
CLS6795611
Corning® 11in x 11in stirrer, model PC-611
AC/DC input 120 V AC, plate W × L 11 in. × 11 in.
Synonym(s):
magnetic stirrer, magnetic stirrers, stirrers
About This Item
UNSPSC Code:
41103803
NACRES:
NB.73
Pricing and availability is not currently available.
manufacturer/tradename
Corning 6795-611
availability
not available in EU
stirring range measuring range
100-1500 rpm
plate W × L
11 in. × 11 in.
General description
These Corning PC-611 stirrers have an 11 x 11 inch plastic (ABS) top and can stir large volumes of liquid with ease. These stirrers are designed for 5 gallon vessels (19L), but can stir up to 10 gallons and can handle many applications that previously required an overhead stirrer. This powerful stirring is achieved with a very compact design that conserves bench space. The stirrer uses a special rare earth magnet that is constructed out of for exceptional coupling strength. The magnet′s strength also centers the stir bar quickly inside the vessel. They operate on 120V/60Hz and are UL and CUL approved to meet needs in North America and other locations with similar requirements.
Heavy duty motor and rugged plastic (ABS) top will provide years of reliable service. -More compact and lower cost than competitive units. -UL/CUL approved. -1 year warranty.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)