Skip to Content
Merck
All Photos(1)

Key Documents

CLS615002L

PYREXPLUS® narrow-mouth reagent bottle, with Pyrex® ST stopper, protective coating

capacity 2000 mL

Synonym(s):

Corning® 61500

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
R$1,088.00

R$1,088.00


Check Cart for Availability


Select a Size

Change View
1 EA
R$1,088.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

R$1,088.00


Check Cart for Availability

material

borosilicate glass bottle
clear bottle
cylindrical bottle
narrow-mouth bottle

stopper size

29

feature

autoclavable
cap

manufacturer/tradename

Corning 61500-2L

capacity

2000 mL

diam. × H

113 mm × 248 mm

Looking for similar products? Visit Product Comparison Guide

General description

Features a protective plastic coating that helps prevent glass from shattering and reduces any spills. Fully autoclavable (121°C) and resistant to thermal shock. Bottle has a ground neck,

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
PYREXPLUS is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service