Skip to Content
Merck
All Photos(1)

Documents

CLS5563525

Pyrex® volumetric flask, low actinic, heavy duty wide mouth, with Pyrex stopper

capacity 25 mL

Synonym(s):

Corning® 55635

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121804
NACRES:
NB.43

stopper size

13

feature

class A

packaging

case of 6 ea

manufacturer/tradename

Corning 55635-25

capacity

25 mL

Looking for similar products? Visit Product Comparison Guide

General description

Low actinic stained glass provides protection for materials sensitive to light. the protective color is an intergral part of the flask, which retains the mechanical strength, chemical stability and thermal resistance of the Pyrex brand labware.

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service