CLS53201L
Pyrex® heavy-wall graduated filtering flask
capacity 1,000
Synonym(s):
conical flasks, filter flasks, filtering flask, filtering flasks, flask filter, Corning® 5320
Select a Size
R$4,206.00
Select a Size
About This Item
R$4,206.00
Recommended Products
material
borosilicate glass
flat bottom flask
narrow-mouth
Joint
joint stopper8
description
without hose connection
sterility
non-sterile
feature
closure type Rubber stopper
packaging
case of 12
pack of 6
manufacturer/tradename
Corning 5320-1L
volume measuring range
300-1,000 mL
capacity
1,000
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)