Key Documents
CLS498010
Pyrex® narrow-mouth graduated Erlenmeyer flask
capacity 10 mL, stopper joint: 00
Synonym(s):
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks glass, Corning® 4980
Select a Size
12 EA
R$1,290.00
R$1,290.00
Check Cart for Availability
Select a Size
Change View
12 EA
R$1,290.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
R$1,290.00
Check Cart for Availability
material
borosilicate glass
narrow-mouth
Joint
joint: stopper00
sterility
non-sterile
feature
cap: no
packaging
case of 12
manufacturer/tradename
Corning 4980-10
availability
not available in EU
capacity
10 mL
General description
  • Pyrex® narrow-mouth graduated Erlenmeyer flasks are made of borosilicate glass for optical clarity and mechanical strength.
  • Flasks have durable white enamel graduations and an extra-large marking spot.
  • Features an improved tooled heavy-duty top finish.
Approximate graduations are in durable white enamel. An extra large marking space is also provided. Heavy, durable top tooling.
Other Notes
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)