Key Documents
CLS4965100
Pyrex® round-bottom three neck angle type distilling flask
capacity 100 mL
Synonym(s):
bottom flask, flask round, flask round bottom
Select a Size
4 EA
R$4,898.00
R$4,898.00
Available to ship onApril 17, 2025Details
Select a Size
Change View
4 EA
R$4,898.00
About This Item
UNSPSC Code:
41104812
R$4,898.00
Available to ship onApril 17, 2025Details
material
borosilicate glass
round bottom flask
three-neck (Vertical)
Joint
center joint ST/NS24/40
side joint ST/NS19/38
sterility
non-sterile
feature
closure type Standard Taper joint
packaging
pack of 1
case of 4
manufacturer/tradename
Corning 4965-100
capacity
100 mL
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)