Key Documents
CLS4960500
Pyrex® round-bottom three neck vertical unequal height flask
capacity 500 mL, center joint: ST/NS 34/45
Synonym(s):
bottom flask, flask round, flask round bottom
Select a Size
1 EA
R$2,541.00
4 EA
R$4,010.00
R$2,541.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$2,541.00
4 EA
R$4,010.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
R$2,541.00
Check Cart for Availability
material
borosilicate glass
round bottom flask
three-neck (Vertical)
Joint
center joint: ST/NS34/45
side joint ST/NS24/40
sterility
non-sterile
feature
closure type Standard Taper joint
packaging
pack of 1
case of 4
manufacturer/tradename
Corning 4960-500
capacity
500 mL
neck size
(1)
General description
With full length joints.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)