Key Documents
CLS4320C5L
Pyrex® round-bottom heavy wall boiling flask, short neck with joint
capacity 5,000 mL, joint: ST/NS 45/50
Synonym(s):
bottom flask, flask round, round flask, Corning® 4320C
Select a Size
1 EA
R$1,465.00
R$1,465.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$1,465.00
About This Item
UNSPSC Code:
41121804
R$1,465.00
Check Cart for Availability
material
borosilicate glass
round bottom flask
short
Joint
joint: ST/NS45/50
sterility
non-sterile
feature
closure type Standard taper joint
packaging
case of 1
manufacturer/tradename
Corning 4320C-5L
capacity
5,000 mL
neck size
(1)
General description
Pyrex heavy duty boiling flask provides increased mechanical strength. The flask features walls that are 25-30% thicker than the standard version.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)