CLS36060600F
Pyrex® Buchner funnel with fritted disc
capacity 600 mL, porosity 4-5.5 μm
Synonym(s):
Corning® 36060
Select a Size
R$683.00
Select a Size
About This Item
R$683.00
Recommended Products
manufacturer/tradename
Corning 36060-600F
capacity
600 mL
disc diam.
90 mm
pore size
4-5.5 μm porosity
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)