Skip to Content
Merck
All Photos(1)

Key Documents

CF0043

Sigma-Aldrich

3,5-Bis(trifluoromethyl)benzyl azide

Synonym(s):

1-(Azidomethyl)-3,5-bis(trifluoromethyl)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5F6N3
CAS Number:
Molecular Weight:
269.15
MDL number:
UNSPSC Code:
12352125
NACRES:
NA.06

Quality Level

SMILES string

[N-]=[N+]=NCC1=CC(C(F)(F)F)=CC(C(F)(F)F)=C1

InChI

1S/C9H5F6N3/c10-8(11,12)6-1-5(4-17-18-16)2-7(3-6)9(13,14)15/h1-3H,4H2

InChI key

JKGHUBCPWOLOFQ-UHFFFAOYSA-N

General description

3,5-Bis(trifluoromethyl)benzyl azide is a fluoroalkyl azide reagent active in copper-catalyzed azide-alkyne cycloaddition (click reaction). In addition to its use as a highly fluorinated building block, it can also be functionalized onto biomolecules, advanced materials, or polymers containing an alkyne. Its relative stability and number of magnetically equivalent fluorines make it a highly effective 19F NMR sensor for these and other materials.

Learn more about fluorinated click chemistry

Automate your fluorination reactions with Synple Automated Synthesis Platform (SYNPLE-SC002)

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

195.8 °F

Flash Point(C)

91 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Fluorinated azides are a new tool that can be used to introduce fluorinated groups to molecules. These molecules can utilize click chemistry to easily add fluorinated groups.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service