Skip to Content
Merck
All Photos(1)

Key Documents

CDS001465

Sigma-Aldrich

6-Methyl-3,5-heptadien-2-one

AldrichCPR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12O
Molecular Weight:
124.18
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

liquid

SMILES string

C\C(C)=C\C=C\C(C)=O

InChI

1S/C8H12O/c1-7(2)5-4-6-8(3)9/h4-6H,1-3H3/b6-4+

InChI key

KSKXSFZGARKWOW-GQCTYLIASA-N

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

177.1 °F - closed cup

Flash Point(C)

80.6 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marlin E Rice et al.
Journal of economic entomology, 113(5), 2269-2275 (2020-07-23)
The wood-boring larvae of longhorned beetles (Coleoptera: Cerambycidae) can be important pests of woody plants, particularly as invasive species introduced by international commerce. Previous research has revealed that cerambycid species native to different parts of the world often share the
De-Wei Chen et al.
Food chemistry, 286, 71-77 (2019-03-05)
Lipids, particularly phospholipids, are known to play a significant role in the characteristic aroma of the different meat species. Both neutral lipids and phospholipids were extracted from egg yolk and added to minced chicken (1% w/w) prior to cooking in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service