Key Documents
CAD6184
1-way tubing connector
FLL to Tuohy Borst (1-5 French) (plated brass)
Select a Size
10 EA
R$5,476.00
R$5,476.00
Check Cart for Availability
Select a Size
Change View
10 EA
R$5,476.00
About This Item
UNSPSC Code:
40183110
NACRES:
NB.03
R$5,476.00
Check Cart for Availability
packaging
pack of 10 ea
manufacturer/tradename
Cadence Science 6184
General description
  • Plated brass or 316 stainless steel
  • Meets ISO 594 specifications
  • Final assembly water tested at 45 PSI
Female Luer Lock (FLL) - contains a tabbed hub for secure fitting to a threaded male Luer lock.
Micro-Mate® Female Luer Lock (MMFLL) - designed and manufactured to reduce the stripping of threads which occurs when metal adapters are mated with plastic male Luer lock adapters. Uniquely designed to facilitate easy cleaning and maintenance.
Male Luer Lock (MLL) - male Luer slip with a threaded sheath for a secure fitting to a female Luer lock.
Male Luer (ML) - male Luer slip that fits with a female Luer.
Legal Information
Micro-Mate is a registered trademark of Cadence Science
Choose from one of the most recent versions:
Certificates of Analysis (COA)