Key Documents
CAD4170
Stainless steel 304 syringe needle
gauge 25, L 1 in.
Select a Size
12 EA
R$3,121.00
R$3,121.00
Check Cart for Availability
Select a Size
Change View
12 EA
R$3,121.00
About This Item
UNSPSC Code:
42142600
NACRES:
NB.03
R$3,121.00
Check Cart for Availability
description
Standard Hub with 304 Stainless Steel Tubing
packaging
pack of 12 ea
manufacturer/tradename
Cadence Science 4170
L
1 in.
gauge
25
General description
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
Choose from one of the most recent versions:
Certificates of Analysis (COA)