Skip to Content
Merck
All Photos(1)

Key Documents

C71009

Sigma-Aldrich

3-Chlorostyrene

98%, contains 0.1% 3,5-di-tert-butylcatechol as stabilizer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHC6H4Cl
CAS Number:
Molecular Weight:
138.59
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

liquid

contains

0.1% 3,5-di-tert-butylcatechol as stabilizer

refractive index

n20/D 1.562 (lit.)

density

1.09 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Clc1cccc(C=C)c1

InChI

1S/C8H7Cl/c1-2-7-4-3-5-8(9)6-7/h2-6H,1H2

InChI key

BOVQCIDBZXNFEJ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leire Ruiz-Rubio et al.
Materials (Basel, Switzerland), 12(1) (2019-01-10)
Polymeric thin films patterned with honeycomb structures were prepared from poly(x-chlorostyrene) and statistical poly(x-chlorostyrene-co-styrene) copolymers by static breath figure method. Each polymeric sample was synthesized by free radical polymerization and its solution in tetrahydrofuran cast on glass wafers under 90%

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service